4-cyanoquinoline 97%

Supplier: Ambeed
Warning

Synonyms: Quinoline-4-carbonitrile

A123878-100MG A123878-1G A123878-250MG A123878-5G
77285-030EA 23.69 USD
77285-030 77285-032 77285-034 77747-893
4-cyanoquinoline 97%
4-cyanoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₆N₂
MW: 154.168 g/mol
MDL Number: MFCD00972111
CAS Number: 2973-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR