4-Bromo-3-chlorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A123846-100G
A123846-10G
A123846-1G
A123846-25G
A123846-5G
77285-020EA
101.97
USD
77285-020
77285-022
77285-024
77285-026
77285-028
4-Bromo-3-chlorobenzoic acid 98%
4-Bromo-3-chlorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄BrClO₂ MW: 235.46 g/mol |
MDL Number:
MFCD08276864 CAS Number: 25118-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...