2,4-Dichloro-5-methoxypyrimidine 98%

Supplier: Ambeed
Warning

A123815-100G A123815-10G A123815-1G A123815-25G A123815-5G
77284-998EA 108.15 USD
77284-998 77285-000 77285-002 77285-004 77285-006
2,4-Dichloro-5-methoxypyrimidine 98%
2,4-Dichloro-5-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂O
MW: 179.00 g/mol
MDL Number: MFCD02685670
CAS Number: 19646-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR