FLI-06 98%

Supplier: Ambeed
Warning

Synonyms: Cyclohexyl 1,4,5,6,7,8-hexahydro-2,7,7-trimethyl-4-(4-nitrophenyl)-5-oxo-3-quinolinecarboxylate, Cyclohexyl 2,7,7-trimethyl-4-(4-nitrophenyl)-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxylate

A123790-10MG A123790-1MG A123790-50MG A123790-5MG
77234-246EA 161.71 USD
77234-246 77234-248 77234-250 77234-252
FLI-06 98%
FLI-06

Precaución: for Research Use Only

Formula: C₂₅H₃₀N₂O₅
MW: 438.52 g/mol
MDL Number: MFCD00330996
CAS Number: 313967-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR