4-Fluoropiperidine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A123750-1G
A123750-250MG
A123750-25G
A123750-5G
77284-992EA
9
USD
77284-992
77284-994
77284-996
77566-366
4-Fluoropiperidine hydrochloride 97%
4-Fluoropiperidine hydrochloride
Precaución: For research use only.
Formula:
C₅H₁₁ClFN |
MDL Number:
MFCD03452786 CAS Number: 57395-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...