3-[2-(Ethoxycarbonyl)ethyl]benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-(3-Ethoxy-3-oxopropyl)phenyl)boronic acid

A123701-100MG A123701-1G A123701-250MG A123701-5G
77271-678EA 29 USD
77271-678 77271-680 77271-682 77271-684
3-[2-(Ethoxycarbonyl)ethyl]benzeneboronic acid 98%
3-[2-(Ethoxycarbonyl)ethyl]benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₄
MDL Number: MFCD08689511
CAS Number: 913835-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR