Isonipecotamide 95%

Supplier: Ambeed
Warning

A123635-100G A123635-10G A123635-25G A123635-500G A123635-5G
77271-668EA 27 USD
77271-668 77271-670 77271-672 77271-674 77271-676
Isonipecotamide 95%
Isonipecotamide

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MDL Number: MFCD00038012
CAS Number: 39546-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR