1-(2-Methoxyethoxy)-4-nitrobenzene 98%

Supplier: Ambeed
Warning

A123582-1G A123582-250MG A123582-25G A123582-5G
77284-974EA 17 USD
77284-974 77284-976 77284-978 77284-980
1-(2-Methoxyethoxy)-4-nitrobenzene 98%
1-(2-Methoxyethoxy)-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄
MDL Number: MFCD00463497
CAS Number: 22483-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR