3-Hydroxy-3-methylglutaric acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-3-methylpentanedioic acid, Meglutol

A123578-1G A123578-5G A123578-100MG A123578-250MG
77271-664EA 164.8 USD
77271-664 77271-666 77571-434 77747-892
3-Hydroxy-3-methylglutaric acid 98%
3-Hydroxy-3-methylglutaric acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₅
MW: 162.14 g/mol
MDL Number: MFCD00002712
CAS Number: 503-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR