4,5-Dichloro-3(2H)-pyridazinone 97%
Supplier: Ambeed
|
Warning
|
A123395-1000G
A123395-100G
A123395-10G
A123395-25G
A123395-500G
A123395-5G
77284-958EA
217
USD
77284-958
77284-960
77284-962
77284-964
77284-966
77284-968
4,5-Dichloro-3(2H)-pyridazinone 97%
4,5-Dichloro-3(2H)-pyridazinone
Precaución: for Research Use Only
Formula:
C₄H₂Cl₂N₂O |
MDL Number:
MFCD00051504 CAS Number: 932-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...