1,2,3,4-Tetrahydro-1-methylpyrrolo[1,2-a]pyrazine 97%

Supplier: Ambeed
Warning

A123327-10G A123327-1G A123327-250MG A123327-5G
77234-214EA 48.41 USD
77234-214 77234-216 77234-218 77234-220
1,2,3,4-Tetrahydro-1-methylpyrrolo[1,2-a]pyrazine 97%
1,2,3,4-Tetrahydro-1-methylpyrrolo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MW: 136.1900 g/mol
MDL Number: MFCD00296984
CAS Number: 73627-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR