4-Hydrazinyl-N,N-dimethylaniline dihydrochloride 95%

Supplier: Ambeed
Warning

A123272-100MG A123272-250MG
77271-642EA 17 USD
77271-642 77271-644
4-Hydrazinyl-N,N-dimethylaniline dihydrochloride 95%
4-Hydrazinyl-N,N-dimethylaniline dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅Cl₂N₃
MW: 224.13 g/mol
MDL Number: MFCD31619107
CAS Number: 27076-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR