(1-Methylpiperidin-4-yl)methanamine 97%

Supplier: Ambeed
Danger

A123245-10G A123245-1G A123245-5G
77271-636EA 94 USD
77271-636 77271-638 77271-640
(1-Methylpiperidin-4-yl)methanamine 97%
(1-Methylpiperidin-4-yl)methanamine

Precaución: for Research Use Only

Formula: C₇H₁₆N₂
MDL Number: MFCD05022430
CAS Number: 7149-42-0
UN: 2733
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR