2,4,7-Trichloroquinazoline 98%

Supplier: Ambeed
Warning

A123158-1G A123158-250MG
77246-184EA 38.11 USD
77246-184 77246-186
2,4,7-Trichloroquinazoline 98%
2,4,7-Trichloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₃Cl₃N₂
MW: 233.4818 g/mol
MDL Number: MFCD09954887
CAS Number: 6625-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR