3,5-Dichloro-2-hydroxybenzonitrile 97%

Supplier: Ambeed
Warning

A122976-100MG A122976-250MG
77271-616EA 321.36 USD
77271-616 77271-618
3,5-Dichloro-2-hydroxybenzonitrile 97%
3,5-Dichloro-2-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO
MW: 188.01 g/mol
MDL Number: MFCD02261938
CAS Number: 3336-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR