4-(Trifluoromethyl)indole 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)-1H-indole

A122886-100MG A122886-1G A122886-250MG
77284-936EA 27.81 USD
77284-936 77284-938 77284-940
4-(Trifluoromethyl)indole 98%
4-(Trifluoromethyl)indole

Precaución: for Research Use Only

Formula: C₉H₆F₃N
MW: 185.149 g/mol
MDL Number: MFCD09954766
CAS Number: 128562-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR