6-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)hexanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)hexanoic acid

A122680-250MG A122680-1G A122680-5G
77258-144EA 115.36 USD
77258-144 77614-962 77747-887
6-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)hexanoic acid 97%
6-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)hexanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₄
MW: 367.44 g/mol
MDL Number: MFCD30596881
CAS Number: 173690-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR