4-Amylcyclohexanol (cis- and trans- mixture) 98%

Supplier: Ambeed
Warning

Synonyms: 4-Pentylcyclohexanol

A122652-100G A122652-10G A122652-5G
77284-916EA 180 USD
77284-916 77284-918 77284-920
4-Amylcyclohexanol (cis- and trans- mixture) 98%
4-Amylcyclohexanol (cis- and trans- mixture)

Precaución: for Research Use Only

Formula: C₁₁H₂₂O
MDL Number: MFCD01017412
CAS Number: 54410-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR