4-(2-Chloroethoxy)benzaldehyde 98%

Supplier: Ambeed
Warning

A122639-1G A122639-5G
77271-594EA 13 USD
77271-594 77747-886
4-(2-Chloroethoxy)benzaldehyde 98%
4-(2-Chloroethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MDL Number: MFCD00043699
CAS Number: 54373-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR