Sodium 4-(nicotinamido)butanoate 97%

Supplier: Ambeed
Warning

A122628-100G A122628-10G A122628-25G A122628-5G
77284-902EA 45 USD
77284-902 77284-904 77284-906 77284-908
Sodium 4-(nicotinamido)butanoate 97%
Formula: C₁₀H₁₁N₂NaO₃
MDL Number: MFCD00887262
CAS Number: 62936-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR