Trifluoromethanesulfonamide 95%

Supplier: Ambeed
Warning

A122541-10G A122541-1G A122541-250MG A122541-25G A122541-5G A122541-500G
77271-584EA 24 USD
77271-584 77271-586 77271-588 77271-590 77271-592 77571-368
Trifluoromethanesulfonamide 95%
Trifluoromethanesulphonamide

Precaución: for Research Use Only

Formula: CH₂F₃NO₂S
MDL Number: MFCD00068714
CAS Number: 421-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR