N-(2,2-Dimethoxyethyl)-2-phenylacetamide 95%
Supplier: Ambeed
|
Warning
|
A122496-10G
A122496-25G
A122496-5G
77234-176EA
28.84
USD
77234-176
77234-178
77234-180
N-(2,2-Dimethoxyethyl)-2-phenylacetamide 95%
N-(2,2-Dimethoxyethyl)-2-phenylacetamide
Precaución: for Research Use Only
Formula:
C₁₂H₁₇NO₃ MW: 223.26 g/mol |
MDL Number:
MFCD17257378 CAS Number: 89314-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...