P,P'-(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis[N,N,N',N'-tetraethyl-phosphonous diamide] 98%

Supplier: Ambeed
Warning

A122476-100MG A122476-250MG A122476-5G A122476-1G
77246-168EA 25.75 USD
77246-168 77246-170 77246-172 77566-308
P,P'-(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis[N,N,N',N'-tetraethyl-phosphonous diamide] 98%
P,P'-(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis[N,N,N',N'-tetraethyl-phosphonous diamide]

Precaución: For research use only.

Formula: C₃₁H₅₂N₄OP₂
MW: 558.71 g/mol
MDL Number: MFCD13182459
CAS Number: 349100-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR