(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: H-Leu-OMe·HCl, H-Leu-OMe.HCl, L-Leucine methyl ester hydrochloride

A122432-1000G A122432-100G A122432-10G A122432-25G A122432-500G A122432-5G
77271-572EA 117 USD
77271-572 77271-574 77271-576 77271-578 77271-580 77271-582
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 96%
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂·HCl
MW: 181.7 g/mol
MDL Number: MFCD00012494
CAS Number: 7517-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR