2,5-Difluorophenylhydrazinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,5-Difluorophenyl)hydrazine hydrochloride, 2,5-Difluorophenylhydrazine hydrochloride

A122395-100G A122395-10G A122395-1G A122395-25G A122395-5G
77284-880EA 275.01 USD
77284-880 77284-882 77284-884 77284-886 77284-888
2,5-Difluorophenylhydrazinium chloride 98%
2,5-Difluorophenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₆H₇ClF₂N₂
MW: 180.58 g/mol
MDL Number: MFCD00013385
CAS Number: 175135-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR