5-Fluoro-2-nitrophenol 98%

Supplier: Ambeed
Danger

Synonyms: 4-Fluoro-2-hydroxynitrobenzene

A122359-25G A122359-500G A122359-5G A122359-100G
77284-874EA 11 USD
77284-874 77284-876 77284-878 77571-354
5-Fluoro-2-nitrophenol 98%
5-Fluoro-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄FNO₃
MW: 157.1 g/mol
Boiling Pt: 260
MDL Number: MFCD00007107
CAS Number: 446-36-6
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR