Ethyl 3-(4-(methylamino)-3-nitro-N-(pyridin-2-yl)benzamido)propanoate 97%

Supplier: Ambeed
Warning

A122175-100G A122175-25G A122175-500G A122175-5G
77234-158EA 66 USD
77234-158 77234-160 77234-162 77234-164
Ethyl 3-(4-(methylamino)-3-nitro-N-(pyridin-2-yl)benzamido)propanoate 97%
Ethyl 3-(4-(methylamino)-3-nitro-N-(pyridin-2-yl)benzamido)propanoate

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₄O₅
MW: 372.37 g/mol
MDL Number: MFCD09833623
CAS Number: 429659-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR