2-Chloro-9-methyl-3H-purin-6(9H)-one 95%

Supplier: Ambeed
Warning

A122166-100MG A122166-1G
77234-156EA 74.16 USD
77234-156 77566-300
2-Chloro-9-methyl-3H-purin-6(9H)-one 95%
2-Chloro-9-methyl-3H-purin-6(9H)-one

Precaución: For research use only.

Formula: C₆H₅ClN₄O
MW: 184.58 g/mol
MDL Number: MFCD28145110
CAS Number: 36323-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR