3-Amino-2-bromo-4,6-dimethylpyridine 95%

Supplier: Ambeed
Warning

A122138-1G A122138-250MG A122138-5G
77271-562EA 24 USD
77271-562 77271-564 77271-566
3-Amino-2-bromo-4,6-dimethylpyridine 95%
3-Amino-2-bromo-4,6-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MDL Number: MFCD03731181
CAS Number: 104829-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR