Methyl-2-amino-3-methylbenzoate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-3-methylbenzoate

A122057-100G A122057-10G A122057-1G A122057-25G A122057-5G
77246-138EA 84.46 USD
77246-138 77246-140 77246-142 77246-144 77246-146
Methyl-2-amino-3-methylbenzoate 96%
Methyl 2-amino-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD06200918
CAS Number: 22223-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR