4-(Methylamino)pyridine 99%

Supplier: Ambeed
Warning

A122030-100G A122030-25G A122030-5G
77271-558EA 331.66 USD
77271-558 77271-560 77566-290
4-(Methylamino)pyridine 99%
4-(Methylamino)pyridine

Precaución: For research use only.

Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00152625
CAS Number: 1121-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR