6-(Cyanomethyl)nicotinonitrile 97%

Supplier: Ambeed
Warning

A121784-100MG A121784-1G A121784-250MG
77234-148EA 85.49 USD
77234-148 77234-150 77234-152
6-(Cyanomethyl)nicotinonitrile 97%
6-(Cyanomethyl)nicotinonitrile

Precaución: for Research Use Only

Formula: C₈H₅N₃
MDL Number: MFCD09924285
CAS Number: 1000516-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR