3-Chloro-4,5-difluorobenzoic acid 96%

Supplier: Ambeed
Warning

A121734-1G A121734-250MG
77271-548EA 37.08 USD
77271-548 77271-550
3-Chloro-4,5-difluorobenzoic acid 96%
3-Chloro-4,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O₂
MW: 192.547 g/mol
MDL Number: MFCD04972707
CAS Number: 150444-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR