2-Amino-4-thiazoleacetic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Aminothiazol-4-yl)acetic acid hydrochloride

A121503-100G A121503-10G A121503-1G A121503-25G A121503-5G
77284-830EA 28.84 USD
77284-830 77284-832 77284-834 77284-836 77284-838
2-Amino-4-thiazoleacetic acid hydrochloride 97%
2-Amino-4-thiazoleacetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂S·HCl
MW: 194.64 g/mol
MDL Number: MFCD06658943
CAS Number: 66659-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR