3-(2-Chloropyrimidin-4-yl)-1-cyclopropyl-1H-indole 97%

Supplier: Ambeed
Warning

A121461-100MG A121461-1G A121461-250MG A121461-5G
77258-122EA 8.24 USD
77258-122 77258-124 77258-126 77258-128
3-(2-Chloropyrimidin-4-yl)-1-cyclopropyl-1H-indole 97%
3-(2-Chloropyrimidin-4-yl)-1-cyclopropyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₅H₁₂ClN₃
MW: 269.72 g/mol
MDL Number: MFCD30723492
CAS Number: 1899922-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR