3,5-Dibromo-1-methylpyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A121430-10G A121430-1G A121430-25G A121430-5G
77271-522EA 38 USD
77271-522 77271-524 77271-526 77271-528
3,5-Dibromo-1-methylpyridin-2(1H)-one 97%
3,5-Dibromo-1-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₅Br₂NO
MDL Number: MFCD00464328
CAS Number: 14529-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR