4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-n-propylbiphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-propylbiphenyl, 4-(Difluoro(3,4,5-trifluorophenoxy)methyl)-3,5-difluoro-4'-propyl-1,1'-biphenyl

A121391-250MG A121391-5G
77234-126EA 6.18 USD
77234-126 77234-128
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-n-propylbiphenyl 97%
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-n-propylbiphenyl

Precaución: for Research Use Only

Formula: C₂₂H₁₅F₇O
MW: 428.35 g/mol
MDL Number: MFCD18451990
CAS Number: 303186-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR