4-Amino-5-chloro-2-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Metoclopramide Impurity C, 4-Amino-5-chloro-o-anisic acid

A121379-100G A121379-1G A121379-25G A121379-5G A121379-500G
77234-118EA 165.83 USD
77234-118 77234-120 77234-122 77234-124 77745-892
4-Amino-5-chloro-2-methoxybenzoic acid 98%
4-Amino-5-chloro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
MDL Number: MFCD00038794
CAS Number: 7206-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR