LW6 97%

Supplier: Ambeed
Warning

Synonyms: CAY10585, 3-[[[4-(Adamantan-1-yl)phenoxy]acetyl]amino]-4-hydroxybenzoic acid methyll ester, Methyl 3-(2-(4-(adamantan-1-yl)phenoxy)acetamido)-4-hydroxybenzoate, Methyl 3-[[[4-(adamantan-1-yl)phenoxy]acetyl]amino]-4-hydroxybenzoate

A121335-100MG A121335-10MG A121335-1MG A121335-25MG A121335-50MG A121335-5MG
77271-510EA 185.4 USD
77271-510 77271-512 77271-514 77271-516 77271-518 77271-520
LW6 97%
LW6

Precaución: for Research Use Only

Formula: C₂₆H₂₉NO₅
MW: 435.51 g/mol
MDL Number: MFCD09907563
CAS Number: 934593-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR