2-((2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)methoxy)-3-hydroxypropyl ((benzyloxy)carbonyl)-L-valinate 97%

Supplier: Ambeed
Warning

Synonyms: N-Carbobenzyloxy-L-valinyl-ganciclovir

A121211-10G A121211-1G A121211-5G
77246-112EA 57 USD
77246-112 77246-114 77246-116
2-((2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)methoxy)-3-hydroxypropyl ((benzyloxy)carbonyl)-L-valinate 97%
2-((2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)methoxy)-3-hydroxypropyl ((benzyloxy)carbonyl)-L-valinate

Precaución: for Research Use Only

Formula: C₂₂H₂₈N₆O₇
MW: 488.49 g/mol
MDL Number: MFCD13194862
CAS Number: 194154-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR