6-(Methoxycarbonyl)picolinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Pyridinedicarboxylic acid monomethyl ester, Dipicolinic acid monomethyl ester, Monomethyl 2,6-Pyridinedicarboxylate, Monomethyl dipicolinate

A120886-100G A120886-10G A120886-1G A120886-25G A120886-5G
77234-104EA 218 USD
77234-104 77234-106 77234-108 77234-110 77234-112
6-(Methoxycarbonyl)picolinic acid 97%
6-(Methoxycarbonyl)picolinic acid

Precaución: for Research Use Only

Formula: C8H7NO4
MW: 181.1455 g/mol
MDL Number: MFCD00806109
CAS Number: 7170-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR