1-Chloro-2-fluoro-4-nitrobenzene 98%

Supplier: Ambeed
Warning

A120824-5G A120824-10G A120824-25G
77284-808EA 10 USD
77284-808 77571-244 77747-878
1-Chloro-2-fluoro-4-nitrobenzene 98%
1-Chloro-2-fluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MDL Number: MFCD00215833
CAS Number: 350-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR