N-(3-Ethynylphenyl)-7,8,10,11,13,14-hexahydro-[1,4,7,10]tetraoxacyclododecino[2,3-g]quinazolin-4-amine 98+%

Supplier: Ambeed
Warning

A120776-10MG A120776-1MG A120776-50MG A120776-5MG
77234-090EA 37.08 USD
77234-090 77234-092 77234-094 77234-096
N-(3-Ethynylphenyl)-7,8,10,11,13,14-hexahydro-[1,4,7,10]tetraoxacyclododecino[2,3-g]quinazolin-4-amine 98+%
N-(3-Ethynylphenyl)-7,8,10,11,13,14-hexahydro-[1,4,7,10]tetraoxacyclododecino[2,3-g]quinazolin-4-amine

Precaución: for Research Use Only

Formula: C₂₂H₂₁N₃O₄
MW: 391.42 g/mol
MDL Number: MFCD22124501
CAS Number: 610798-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR