3,5-Di-tert-butylaniline 95%

Supplier: Ambeed
Warning

A120608-100MG A120608-1G A120608-250MG A120608-5G A120608-25G
77271-480EA 12.36 USD
77271-480 77271-482 77271-484 77271-486 77549-414
3,5-Di-tert-butylaniline 95%
3,5-Di-tert-butylaniline

Precaución: for Research Use Only

Formula: C₁₄H₂₃N
MW: 205.34 g/mol
MDL Number: MFCD00134096
CAS Number: 2380-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR