Methyl 1-[(2'-Cyano-[1,1'-biphenyl]-4-yl)methyl]-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate 98%

Supplier: Ambeed
Warning

A120551-25G A120551-5G
77234-088EA 28 USD
77234-088 77745-887
Methyl 1-[(2'-Cyano-[1,1'-biphenyl]-4-yl)methyl]-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate 98%
Methyl 1-[(2'-Cyano-[1,1'-biphenyl]-4-yl)methyl]-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate

Precaución: for Research Use Only

Formula: C₂₅H₂₁N₃O₃
MDL Number: MFCD09031384
CAS Number: 139481-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR