trans-4-(4-Pentylcyclohexyl)benzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-(trans-4-n-Pentylcyclohexyl)benzonitrile

A120540-100G A120540-10G A120540-1G A120540-250MG A120540-25G A120540-5G
77284-798EA 177.16 USD
77284-798 77284-800 77284-802 77284-804 77284-806 77571-226
trans-4-(4-Pentylcyclohexyl)benzonitrile 97%
trans-4-(4-Pentylcyclohexyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₈H₂₅N
MW: 255.40 g/mol
MDL Number: MFCD00075548
CAS Number: 61204-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR