Methyl-4,6-dihydrothieno[3,4-b]thiophene-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A120523-100MG
A120523-10G
A120523-250MG
A120523-5G
A120523-1G
77234-080EA
5
USD
77234-080
77234-082
77234-084
77234-086
77745-886
Methyl-4,6-dihydrothieno[3,4-b]thiophene-2-carboxylate 95%
Methyl-4,6-dihydrothieno[3,4-b]thiophene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₈O₂S₂ |
MDL Number:
MFCD15096514 CAS Number: 7767-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...