tert-Butyl phenyl ether 96%

Supplier: Ambeed
Warning

Synonyms: tert-Butoxybenzene

A120512-1G A120512-250MG A120512-25G A120512-5G
77234-072EA 14 USD
77234-072 77234-074 77234-076 77234-078
tert-Butyl phenyl ether 96%
tert-Butyl phenyl ether

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MDL Number: MFCD00049018
CAS Number: 6669-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR