4-Fluoro-2-hydroxybenzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-hydroxyphenylboronic acid

A120503-100MG A120503-10G A120503-1G A120503-250MG A120503-5G
77284-788EA 25.75 USD
77284-788 77284-790 77284-792 77284-794 77284-796
4-Fluoro-2-hydroxybenzeneboronic acid 97%
4-Fluoro-2-hydroxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BFO₃
MW: 155.92 g/mol
MDL Number: MFCD04115662
CAS Number: 850568-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR