4-Benzylmorpholine-2-carbonitrile 98%

Supplier: Ambeed
Warning

A120465-1G A120465-25G A120465-5G
77284-782EA 15.45 USD
77284-782 77284-784 77284-786
4-Benzylmorpholine-2-carbonitrile 98%
4-Benzylmorpholine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O
MW: 202.2524 g/mol
MDL Number: MFCD08751344
CAS Number: 126645-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR